[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Drug Delivery](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering/drug-delivery)Solubility Enhancement with Dendrimers
# Solubility Enhancement with Dendrimers
### Read about
- [Introduction](https://www.sigmaaldrich.com#introduction)
- [Experimental](https://www.sigmaaldrich.com#experimental)
- [Results and Discussion](https://www.sigmaaldrich.com#results)
- [Indomethacin](https://www.sigmaaldrich.com#indomethacin)
- [Comparison of Release Profiles](https://www.sigmaaldrich.com#comparison)
- [Conclusion](https://www.sigmaaldrich.com#conclusion)
## [](https://www.sigmaaldrich.com)Introduction
Environmental concerns are driving the replacement of volatile organic solvents by water and aqueous mixtures. This change often creates challenges because many organic molecules show low water solubility. In addition, in some applications, it is necessary to mix hydrophilic and hydrophobic components in one formulation, requiring the use of solubility modifiers. Dendrimers can provide a solution in both scenarios. Dendrimers are core-shell nanostructures with precise architecture and low polydispersity, which are synthesized in a generation-by-generation fashion around a core unit, resulting in a high level of control over size, branching points, and surface functionality (__Figure 1__).1 The ability to tailor dendrimer properties to the needs of a guest molecule makes them ideal carriers for molecular encapsulation, allowing dissolution of hydrophobic materials in water and mixing of hydrophilic and hydrophobic compounds, for example in catalytic applications.2
__Figure 1.__Schematic representation of a G2 dendrimer with 3-fold branching. PAMAM dendrimers have 2-fold branching, resulting in 2x increase in number of surface groups with each generation, G.
In this article, we demonstrate the ability of dendrimers to improve water solubility, using two poorly water-soluble drugs as model materials. Low water solubility and, therefore, poor bioavailability, are a major challenge in drug delivery.3 The anti-cancer drug cisplatin and the non-steroidal antiinflammatory drug indomethacin have been encapsulated into STARBURST® poly(amidoamine) (PAMAM) dendrimers, and their encapsulation efficiency and release profiles have been studied. Compared to control experiments without dendrimers, dendrimer-drug formulations showed up to 37-fold drug solubility enhancement.
## [](https://www.sigmaaldrich.com)Experimental
### Materials
STARBURST® PAMAM dendrimers are products of Dendritic Nanotechnologies, Inc. and currently available through our portfolio. All other reagents and solvents are available and were used as received.
### Encapsulation Protocol
Dendrimers were dissolved in deionized (DI) water and mixed with excess drug. These suspensions were briefly exposed to ultrasonication, and then incubated overnight at 37° C and 100 rpm in a shaking water bath. After equilibration to ambient temperature, the dendrimer-drug suspensions were filtered through a 0.2-μm nylon syringe filter to remove excess solid drug. Non-encapsulated drug was removed by dialysis using a 1-kDa cutoff membrane against DI water at 4° C for 30 min. Drug samples were freeze-dried and analyzed for encapsulated drug content by UV spectroscopy at 320 nm using a Perkin Elmer™ Lambda 2 UV/VIS Spectrophotometer (indomethacin) or by inductively coupled plasma (ICP) spectroscopy (cisplatin). Procedure was performed in triplicate.
### *In Vitro* Release Protocol
Dendrimer-drug conjugates were re-dissolved in DI water or saline solution and placed in a dialysis bag using a 1-kDa cutoff membrane at 37° C. At certain time intervals, aliquots were removed from the outer dialysis compartment, freeze-dried and analyzed by UV spectroscopy or ICP as described above.
## [](https://www.sigmaaldrich.com)Results and Discussion
### Cisplatin
Encapsulation of cisplatin is independent of dendrimer core and generation as evident from __Table 1__ (P > 0.05). The effect of drug/dendrimer molar ratios on the percentage of Pt content was studied using dendrimer G3.5 with diaminoethane (EDA) core and sodium carboxylate surface groups. Increasing the cisplatin/dendrimer molar ratio resulted in enhanced Pt content of the dendrimer formulations (__Table 2__). The effect of dendrimer-to-cisplatin loading on *in vitro* release was studied using dendrimer G3.5 (__Figure 2__). The release rate was correlated to the amount of drug loaded into the dendrimers, with fast release at high loading but slow release (long retention time) at low loading. Long retention is desirable in technical, catalytic applications. *In vitro* release of cisplatin was independent of the dendrimer core size. The release profiles in saline solution (high ionic strength) were similar to the release profiles in DI water (data not shown).
Dendrimer
CoreDendrimer
GenPt content (%w/v)Mean±SDProd. No.
EDAa3.527.27 ± 0.67[412430](https://www.sigmaaldrich.com/US/en/product/ALDRICH/412430)
EDA4.527.6 ± 1.21[470457](https://www.sigmaaldrich.com/US/en/product/ALDRICH/470457)
adiaminoethane; bdiaminobutane; cdiaminododecane
Table 1Effect of core and generation on cisplatin loading of dendrimers.
| | |
|----------------------------|-------------------|
| Drug/dendrimer molar ratio | Pt content (%w/v) |
| 9.4 | 10.92 |
| 19 | 18.65 |
| 28 | 26.19 |
| 53 | 35.8 |
| 78 | 42.8 |
| 106 | 50.1 |
| 304 | 58.05 |
Table 2The effect of drug/dendrimer molar ratio on the percentage of Pt content (STARBURST® dendrimer G3.5 with EDA core and COONa surface).
__Figure 2.__In vitro release profiles of cisplatin formulated with STARBURST® dendrimer G3.5 sodium carboxylate surface in DI water at different molar ratios. Ratio listed next to formulation denotes number of drug molecules associated with dendrimer.
## [](https://www.sigmaaldrich.com)Indomethacin
STARBURST® dendrimers of different generations (G1-6) and surface groups have been used to study encapsulation and release of indomethacin (__Figure 3__). Water solubility of indomethacin was dramatically enhanced through encapsulation, depending on dendrimer size (G6 > G5 > G4 > G3) and surface groups (NH2 > OH > COO¯). *In vitro* release of indomethacin showed a similar but inverted relationship to size (G3 > G4 > G5 > G6) and surface groups (NH2 < OH < COO¯) (Figure 3). Lag time and very slow release from STARBURST® dendrimer G4 with amine surface groups is a result of strong electrostatic interactions and hydrogen bonding of indomethacin between dendrimer surface and the carboxylate group of the drug. Fast release from STARBURST® dendrimer G4 with sodium carboxylate surface groups reflects electrostatic repulsion between adjacent surface groups. This repulsion opens the dendritic structure and reduces its container properties, as well as repulsion between drug molecules and dendrimer surface. However, drug release can be slowed down by modification of the dendrimer surface, e.g., by surface conjugation of poly(ethylene glycol) chains such as PEG-500. Such PEG-modified dendrimers are now commercially available (See PEG dendrimer kit)
, carbomethoxypyrrolidinone, amidoethanol, sodium carboxylate, succinamic acid, and tris(hydroxymethyl)amidomethane.")
__Figure 3.__Encapsulation efficiency of indomethacin into STARBURST® dendrimers. Different dendrimer surfaces shown: primary amine, poly(ethylene glycol), carbomethoxypyrrolidinone, amidoethanol, sodium carboxylate, succinamic acid, and tris(hydroxymethyl)amidomethane.
__Figure 4.__Indomethacin release from STARBURST® dendrimers G4 with primary amine and sodium carboxylate surface groups
## [](https://www.sigmaaldrich.com)Comparison of Release Profiles
Release of the drug from the dendrimer matrix depends upon the nature of the drug and its interactions with the dendrimer. For example, the repulsive interaction between the dendritic carrier and indomethacin mentioned above results in fast release of this drug from PAMAM® dendrimers G4 with carboxylate surface groups compared to cisplatin (56.9% *versus* 25.3% after 5 h). Consequently, release profiles can be tailored to the application needs by choice of the dendrimer surface groups (e.g., amine versus carboxylate) as well as the drug/dendrimer molar ratio (__Figures 2 and 4__). Judicious selection of the dendrimer therefore can control the release profile of the encapsulated material.
## [](https://www.sigmaaldrich.com)Conclusion
Formulation of poorly water-soluble drugs with STARBURST® PAMAM dendrimers resulted in increased water solubility, and therefore, enhanced bioavailability of these drugs. This behavior has been demonstrated for two drug classes, small metal ion containing drugs such as cisplatin, and organic molecules such as indomethacin. However, these properties of dendrimers are not restricted to drug molecules but can also be applied to catalysts and organic molecules, where dendrimers function as solubility enhancer and modifier, allowing to mix otherwise immiscible materials.[2](https://www.sigmaaldrich.com#ref) Dendritic Nanotechnologies’ new Priostar™ dendrimer family provides these carriers at low cost and enhanced thermal and hydrolytic stability compared to STARBURST® PAMAM dendrimers, making them ideal candidates for commercializing industrial applications.
## Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Tomalia D. Aldrichimica Acta. AN. 37(1-2):39-48. [https://doi.org/10.4312/an.37.1-2.39-48](https://doi.org/10.4312/an.37.1-2.39-48)
2\.
2007\. Kirk-Othmer Encyclopedia of Chemical Technology. 5th ed..
3\.
Svenson S, Tomalia D. 2005. Dendrimers in biomedical applications - reflections on the field. Advanced Drug Delivery Reviews. 57(15):2106-2129. [https://doi.org/10.1016/j.addr.2005.09.018](https://doi.org/10.1016/j.addr.2005.09.018)
__Related Articles__
- [Photo-Crosslinkable Gelatin Hydrogel: Versatile Materials for (High Resolution) Additive Manufacturing](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/3d-bioprinting/photo-crosslinkable-gelatin-hydrogel)
- [Fluorescent Microparticles and Nanobeads](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/fluorescent-microparticles)
- [Gold Nanoparticles: Properties and Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/gold-nanoparticles)
- [Iron Oxide Nanoparticles: Characteristics & Apps](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/iron-oxide-nanoparticles-characteristics-and-applications)
- [Quantum Dots](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/quantum-dots)
- [Silica-coated Gold Nanoparticles: Properties, Apps](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/silica-coated-gold-nanoparticles)
- [Y-Shape Peg Derivatives in Drug Delivery](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/drug-delivery/applications-of-y-shape-peg-derivatives)
- [Approaches to the design of lipid-based nanocarriers](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/drug-delivery/approaches-to-the-design-of-lipid-based-nanocarriers)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)