Merck
1.17655
HEPA Exhaust filter
new
for use with MAS-100® Sirius®
Note: For More Information on this product, including pricing and availability, please click here!
Pular para
Assistência Técnica
Precisa de ajuda? Nossa equipe de cientistas experientes está à sua disposição.
Estamos aqui para ajudar
Materiais
(Polypropylene)
embalagem
pkg of 1 unit
fabricante/nome comercial
MAS-100® HEPA Exhaust Filter
dilution
(Application - Air monitoring)
compatibilidade
for use with MAS-100® Sirius®
Descrição geral
The MAS-100 Sirius® air sampler features a HEPA exhaust filter classified as an ISO 35 H (H13) filter. This filter is constructed from polypropylene and is embedded in a frame made of polyethylene terephthalate (PET). Its primary function is to prevent the release of contaminants into the cleanroom during the sampling process, particularly when the instrument is transferred from lower to higher-grade cleanrooms.
The Sirius supported by the efficient exhaust HEPA filter operates effectively in both non-controlled areas and cleanroom environments up to grade A (or ISO 5), maintaining compliance with viable and non-viable particle limits. The Exhaust filter is mounted directly at the exhaust of the air path of the MAS-100 Sirius® air sampler. It is recommended to replace the filter on a yearly basis, e.g. during calibration of the air sampler. It can easily be exchanged without the need of any tools.
Aplicação
HEPA exhaust filter in the MAS-100 Sirius® air sampler is essential for maintaining contamination control and high air quality standards in cleanroom environments. It prevents the release of airborne contaminants during the sampling process, ensuring the integrity of the cleanroom is preserved and aiding compliance with stringent regulatory requirements in pharmaceutical, cosmetic, biotechnology, and medical settings.
Informações legais
MAS-100 is a registered trademark of MBV AG
MAS-100 Sirius is a registered trademark of MBV AG
Visite a Biblioteca de Documentos
Conteúdo relacionado
Discover the MAS-100 Sirius® Microbial Air Sampler, designed for reliable air monitoring. Explore features, training, and support services to enhance your lab's compliance and efficiency.