Merck
Antiproliferative effects of CDK4/6 inhibition in CDK4-amplified human liposarcoma in vitro and in vivo.
Molecular cancer therapeutics (2014-07-17)
Yi-Xiang Zhang, Ewa Sicinska, Jeffrey T Czaplinski, Stephen P Remillard, Samuel Moss, Yuchuan Wang, Christopher Brain, Alice Loo, Eric L Snyder, George D Demetri, Sunkyu Kim, Andrew L Kung, Andrew J Wagner
ABSTRAKT
Well-differentiated/dedifferentiated liposarcomas (WD/DDLPS) are among the most common subtypes of soft tissue sarcomas. Conventional systemic chemotherapy has limited efficacy and novel therapeutic strategies are needed to achieve better outcomes for patients. The cyclin-dependent kinase 4 (CDK4) gene is highly amplified in more than 95% of WD/DDLPS. In this study, we explored the role of CDK4 and the effects of NVP-LEE011 (LEE011), a novel selective inhibitor of CDK4/CDK6, on a panel of human liposarcoma cell lines and primary tumor xenografts. We found that both CDK4 knockdown by siRNA and inhibition by LEE011 diminished retinoblastoma (RB) phosphorylation and dramatically decreased liposarcoma cell growth. Cell-cycle analysis demonstrated arrest at G0-G1. siRNA-mediated knockdown of RB rescued the inhibitory effects of LEE011, demonstrating that LEE011 decreased proliferation through RB. Oral administration of LEE011 to mice bearing human liposarcoma xenografts resulted in approximately 50% reduction in tumor (18)F-fluorodeoxyglucose uptake with decreased tumor biomarkers, including RB phosphorylation and bromodeoxyuridine incorporation in vivo. Continued treatment inhibited tumor growth or induced regression without detrimental effects on mouse weight. After prolonged continuous dosing, reestablishment of RB phosphorylation and cell-cycle progression was noted. These findings validate the critical role of CDK4 in maintaining liposarcoma proliferation through its ability to inactivate RB function, and suggest its potential function in the regulation of survival and metabolism of liposarcoma, supporting the rationale for clinical development of LEE011 for the treatment of WD/DDLPS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Gliceryna, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rb1
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%