Merck
  • An integrative process model of enzymatic biodiesel production through ethanol fermentation of brown rice followed by lipase-catalyzed ethanolysis in a water-containing system.
An integrative process model of enzymatic biodiesel production through ethanol fermentation of brown rice followed by lipase-catalyzed ethanolysis in a water-containing system.
Enzyme and microbial technology (2013-01-01)
Daisuke Adachi, Risa Koda, Shinji Hama, Ryosuke Yamada, Kazunori Nakashima, Chiaki Ogino, Akihiko Kondo
ABSTRAKT
We attempted to integrate lipase-catalyzed ethanolysis into fermentative bioethanol production. To produce bioethanol, ethanol fermentation from brown rice was conducted using a tetraploid Saccharomyces cerevisiae expressing α-amylase and glucoamylase. The resultant ethanol was distilled and separated into three fractions with different concentrations of water and fusel alcohols. In ethanolysis using the first fraction with 89.3% ethanol, a recombinant Aspergillus oryzae whole-cell biocatalyst expressing Fusarium heterosporum lipase (r-FHL) afforded the highest ethyl ester content of 94.0% after 96 h. Owing to a high concentration of water in the bioethanol solutions, r-FHL, which works best in the presence of water when processing ethanolysis, was found to be more suitable for the integrative process than a commercial immobilized Candida antarctica lipase. In addition, r-FHL was used for repeated-batch ethanolysis, resulting in an ethyl ester content of more than 80% even after the fifth batch. Fusel alcohols such as 1-butanol and isobutyl alcohol are thought to decrease the lipase activity of r-FHL. Using this process, a high ethyl ester content was obtained by simply mixing bioethanol, plant oil, and lipase with an appropriate adjustment of water concentration. The developed process model, therefore, would contribute to biodiesel production from only biomass-derived feedstocks.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, Isoelectric focusing marker, pI 3.6
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized powder, 30-60 units/mg protein (biuret), ≤0.02% glucose
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, powder, white, ~120 U/mg
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, lyophilized, powder, ~70 U/mg
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ≥260 U/mL, aqueous solution
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Amyloglucosidase from Rhizopus sp., ≥40,000 units/g solid
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)