Merck
[Effect of univalent cations on synthesis of surfactants by Acinetobacter calcoaceticus IMV B-7241].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2013-06-01)
T P Pirog, T A Shevchuk, S I Antoniuk, E Iu Kravchenko, G A Iutinskaia
ABSTRAKT
The effect of univalent cations on activity of key enzymes of C2-metabolism has been investigated in the producer of biosurfactants, Acinetibacter calcoaceticus IMV B-7241 grown on ethanol. It was established that potassium cations are inhibitors of pyroquinolinequinone-dependent alcohol- and acetaldehyde dehydrogenases, the enzymes of biosynthesis of surface-active aminolipids (NADP-dependent glutamate dehydrogenase) and glycolipids (phosphoenopyruvate (PhEP)-carboxikinase), while ammonium cations are activators of these enzymes and PhEP-carboxylase. A decrease of potassium cations concentration in the cultivation medium to 1 mM and increase of the content of amine nitrogen to 10 mM as a result of potassium nitrate substitution by equimolar, as to nitrogen, urea concentration were accompanied by the increase of activity of enzymes of ethanol metabolism and SAS biosynthesis, as well as by the 2-fold increase of conditional concentration of the biosurfactants.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Phosphoenolpyruvate carboxylase microbial, ≥5.0 unit/mg solid
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
L-Glutamic Dehydrogenase (NADP) from Proteus sp., buffered aqueous solution, ≥4,000 units/mL
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%