Merck
Residual bovine serum albumin (BSA) quantitation in vaccines using automated Capillary Western technology.
Analytical biochemistry (2014-05-21)
John W Loughney, Catherine Lancaster, Sha Ha, Richard R Rustandi
ABSTRAKT
Bovine serum albumin (BSA) is a major component of fetal bovine serum (FBS), which is commonly used as a culture medium during vaccine production. Because BSA can cause allergic reactions in humans the World Health Organization (WHO) has set a guidance of 50 ng or less residual BSA per vaccine dose. Vaccine manufacturers are expected to develop sensitive assays to detect residual BSA. Generally, sandwich enzyme-linked immunosorbent assays (ELISA) are used in the industry to detect these low levels of BSA. We report the development of a new improved method for residual BSA detection using the SimpleWestern technology to analyze residual BSA in an attenuated virus vaccine. The method is based on automated Capillary Western and has linearity of two logs, >80% spike recovery (accuracy), intermediate precision of CV <15%, and LOQ of 5.2 ng/ml. The final method was applied to analyze BSA in four lots of bulk vaccine products and was used to monitor BSA clearance during vaccine process purification.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O