Merck
  • Chemical profiling with HPLC-FTMS of exogenous and endogenous chemicals susceptible to the administration of chotosan in an animal model of type 2 diabetes-induced dementia.
Chemical profiling with HPLC-FTMS of exogenous and endogenous chemicals susceptible to the administration of chotosan in an animal model of type 2 diabetes-induced dementia.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Yimin Niu, Feng Li, Chikako Inada, Ken Tanaka, Shiro Watanabe, Hironori Fujiwara, Sachie Sasaki-Hamada, Jun-Ichiro Oka, Kinzo Matsumoto
ABSTRAKT
In our previous study, the daily administration of chotosan (CTS), a Kampo formula consisting of Uncaria and other 10 different crude drugs, ameliorated cognitive deficits in several animal models of dementia including type 2 diabetic db/db mice in a similar manner to tacrine, an acetylcholinesterase inhibitor. The present study investigated the metabonomics of CTS in db/db mice, a type 2 diabetes model, and m/m mice, a non-diabetes control strain, to identify the exogenous and endogenous chemicals susceptible to the administration of CTS using high performance liquid chromatography equipped with an orbitrap hybrid Fourier transform mass spectrometer. The results obtained revealed that the systemic administration of CTS for 20 days led to the distribution of Uncalia plant-derived alkaloids such as rhynchophylline, hirsuteine, and corynoxeine in the plasma and brains of db/db and m/m mice and induced alterations in four major metabolic pathways; i.e., (1) purine, (2) tryptophan, (3) cysteine and methionine, (4) glycerophospholipids in db/db mice. Moreover, glycerophosphocholine (GPC) levels in the plasma and brain were significantly higher in CTS-treated db/db mice than in vehicle-treated control animals. The results of the in vitro experiment using organotypic hippocampal slice cultures demonstrated that GPC (10-30 μM), as well as tacrine, protected hippocampal cells from N-methyl-d-aspartate-induced excitotoxicity in a manner that was reversible with the muscarinic receptor antagonist scopolamine, whereas GPC had no effect on the activity of acetylcholinesterase in vitro. Our results demonstrated that some CTS constituents with neuropharmacological activity were distributed in the plasma and brain tissue following the systemic administration of CTS and may subsequently have affected some metabolic pathways including glycerophospholipid metabolism and cognitive function in db/db mice. Moreover, the present metabonomic analysis suggested that GPC is a putative endogenous chemical that may be involved in the tacrine-like actions of CTS in the present diabetic animal model.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Acetonitrile, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Propidium iodide solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Alkohol metylowy, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Methanol, analytical standard
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Informacje o cenach i dostępności nie są obecnie dostępne.