Merck
In vivo bacteriophage peptide display to tailor pharmacokinetics of biological nanoparticles.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-07-09)
Jessica R Newton-Northup, Marie T Dickerson, Senthil R Kumar, George P Smith, Thomas P Quinn, Susan L Deutscher
ABSTRAKT
Clinical use of most radiolabeled targeting agents has been limited because of the uptake and retention in kidney and/or liver. We hypothesized that bacteriophage (phage) display could be exploited to select for peptide sequences with fast clearance and low kidney uptake with the added ability to redirect phage clearance away from the reticuloendothelial system towards the kidney possessing rapid kidney clearance. In vivo phage display was performed to identify peptides displayed on phage that were excreted rapidly into the urine of mice. A novel in vitro assay using kidney cells, developed to predict in vivo kidney retention, and in vivo pharmacokinetic analyses were performed to characterize selected peptides/phage clones. Forty-three renal clearance clones (RCC) were identified. In vivo mixing experiments and in vitro kidney cell assays identified RCC1-02 as the lead compound. In vivo analysis of fluorescently labeled phage clones demonstrated the ability of RCC1-02 peptide to redirect the biodistribution of the large phage particle towards excretion via the kidney. Pharmacokinetic analysis of [(111)In]-radiolabeled peptides revealed that kidney retention of the control ErBB-2-avid peptide, [(111)In]DOTA-KCCYSL, at 2-h postinjection was 5.7 ± 0.7 %ID/g. In comparison, [(111)In]DOTA-RCC1-02 had kidney retention values of 1.66 ± 0.43 %ID/g, respectively. In vivo phage display can identify phage and corresponding peptides that rapidly clear the renal system. In the future, these peptides may be used to impart favorable pharmacokinetics onto a wide range of radioimaging or therapeutic macromolecules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Wodorowęglan sodu, −40-+140 mesh, ≥95%