Merck
Channel characteristics of VDAC-3 from Arabidopsis thaliana.
Biochemical and biophysical research communications (2015-02-18)
C Berrier, R Peyronnet, J-M Betton, G Ephritikhine, H Barbier-Brygoo, J-M Frachisse, A Ghazi
ABSTRAKT
Four different isoforms of the Voltage-Dependent Anion Channel (VDAC) have been identified in Arabidopsis plant cells. The electrophysiological characteristics of several VDAC channels from animal as well as plant cells are well documented, but those of this model plant are unknown. One isoform, AtVDAC-3 was obtained either directly by cell-free synthesis or produced in Escherichia coli, as inclusion bodies, and re-natured. An electrophysiological study of the purified proteins in planar lipid bilayers showed that both methods yielded proteins with similar channel activity. The characteristics of AtVDAC-3 are that of a bona fide VDAC-like channel.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Urea, European Pharmacopoeia (EP) Reference Standard
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O