Merck
Evaluation of the use of Göttingen minipigs to predict food effects on the oral absorption of drugs in humans.
Journal of pharmaceutical sciences (2014-11-25)
Martin Lau Christiansen, Anette Müllertz, Mats Garmer, Jakob Kristensen, Jette Jacobsen, Bertil Abrahamsson, Rene Holm
ABSTRAKT
This study investigated the oral absorption of drugs in minipigs to predict food effects in man. The protocol was based on a previously described model in dogs and further investigated the food source (i.e., US FDA breakfast or a nutritional drink) and food quantities. Two poorly soluble compounds were investigated [pravastatin (negative food effect) and atazanavir (positive food effect)] in Göttingen minipigs after seven different food regimens. The gastric emptying rate was evaluated by coadministration of acetaminophen. In short, the results demonstrated longer gastric emptying times in minipigs when compared with humans, within a range from 2.3 to 8.4 h dependent on the food regimen. There were no significant differences in drug absorption between fed and fasted state for the two compounds. The study showed that the dog protocol could not be transferred directly to minipigs, but needs further investigation and adjustments in order to get a valid model using Göttingen minipigs for the evaluation of food effects on drug absorption in humans.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Acetaminofen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%