Merck
  • Dopamine D4 receptors linked to protein kinase G are required for changes in dopamine release followed by locomotor activity after repeated cocaine administration.
Dopamine D4 receptors linked to protein kinase G are required for changes in dopamine release followed by locomotor activity after repeated cocaine administration.
Experimental brain research (2015-02-24)
Jeong Hwan Oh, Dong Kun Lee, Yoon-Bo Shim, In Soo Ryu, Su Yeon Seo, Jieun Kim, Ju Hwan Yang, Hyun-Wook Cho, Eun Sang Choe
ABSTRAKT
We previously found that the dopamine D2-type receptors (D2 and D3 receptors), coupled to protein kinase G (PKG), upregulate locomotor activity after repeated cocaine administration. In this study, D4 receptors, another type of D2 receptor also coupled to PKG, were examined to determine their requirement in the regulation of locomotor activity after repeated cocaine administration. The results demonstrated that repeated injections of cocaine (20 mg/kg), given once a day for seven consecutive days, significantly increased extracellular dopamine concentrations. Intra-caudate infusion of the D4 receptor agonist, PD168077 (10 nmol), and the PKG inhibitor, KT5823 (2 nmol), significantly decreased the repeated cocaine-induced increase in dopamine levels and locomotor activity. However, intra-caudate infusion of KT5823, but not PD168077, decreased ∆FosB immunoreactivity elevated by repeated cocaine administration. These findings suggest that D4 receptors linked to PKG could be a key modulator for dopamine release required for changes in locomotor activity caused by repeated cocaine exposure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M