Merck
Subcutaneous phaeohyphomycotic nodule due to Phialemoniopsis hongkongensis sp. nov.
Journal of clinical microbiology (2014-06-27)
Chi-Ching Tsang, Jasper F W Chan, Philip P C Ip, Antonio H Y Ngan, Jonathan H K Chen, Susanna K P Lau, Patrick C Y Woo
ABSTRAKT
Phialemoniopsis species are ubiquitous dematiaceous molds associated with a wide variety of superficial and systemic infections in human. In this study, we isolated a mold from the forearm nodule biopsy specimen from a patient with underlying liver cirrhosis, ankylosing spondylosis, and tuberculosis. He was treated with itraconazole, but unfortunately, he succumbed as a result of disseminated tuberculosis with multiorgan failure. The histology results of the skin biopsy showed necrotizing granulomas in which numerous fungal elements were found. On Sabouraud dextrose agar, the fungal isolate grew as white-to-cream and smooth-to-velvety colonies. Microscopically, oval-to-cylindrical conidia were observed from abundant adelophialides, which possessed barely visible parallel collarettes but no basal septa. The azole drugs voriconazole, itraconazole, and posaconazole, as well as amphotericin B, showed high activities against this fungus. Internal transcribed spacer, 28S nuclear ribosomal DNA (nrDNA), and β-actin and β-tubulin gene sequencing showed that this fungus is most closely related to but distinct from Phialemonium curvata. Matrix-assisted laser desorption ionization-time of flight mass spectrometry (MALDI-TOF MS) and hierarchical cluster analysis showed that the MALDI-TOF MS spectrum of this fungus is most closely related to that of Phialemonium pluriloculosa. We propose a new species, Phialemoniopsis hongkongensis sp. nov., to describe this fungus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent