Merck
  • Diphenyl diselenide and sodium selenite associated with chemotherapy in experimental toxoplasmosis: influence on oxidant/antioxidant biomarkers and cytokine modulation.
Diphenyl diselenide and sodium selenite associated with chemotherapy in experimental toxoplasmosis: influence on oxidant/antioxidant biomarkers and cytokine modulation.
Parasitology (2014-08-12)
Cleber F Barbosa, Alexandre A Tonin, Aleksandro S Da Silva, Maria I De Azevedo, Danieli U Monteiro, Emily P Waczuk, Thiago Duarte, Carine Hermes, Giovana Camillo, Fernanda F Vogel, Luciana Faccio, Paula T Tonin, Patricia Wolkmer, Marta R Leal, Marta M M F Duarte, Rafael N Moresco, Sonia T A Lopes, Mario L De La Rue
ABSTRAKT
SUMMARY The aim of this study was to assess the effect of sulfamethoxazole/trimethoprim (ST) supplemented with diphenyl diselenide and sodium selenite in experimental toxoplasmosis, on oxidant/antioxidant biomarkers and cytokine levels. Eighty-four BALB/c mice were divided in seven groups: group A (negative control), and groups B to G (infected). Blood and liver samples were collected on days 4 and 20 post infection (p.i.). Levels of thiobarbituric acid (TBA) reactive substances and advanced oxidation protein products (AOPP) were assessed in liver samples. Both biomarkers were significantly increased in infected groups on day 4 p.i., while they were reduced on day 20 p.i., compared with group A. Glutathione reductase (GR) activity significantly (P<0·01) increased on day 4 p.i., in group G, compared with group A. INF-γ was significantly increased (P<0·001) in both periods, day 4 (groups B, C, F and G) and 20 p.i. (groups C, F and G). IL-10 significantly reduced (P<0·001) on day 4 p.i. in group B; however, in the same period, it was increased (P<0·001) in groups C and G, compared with group A. On day 20 p.i., IL-10 increased (P<0·001) in groups F and G. Therefore, our results highlighted that these forms of selenium, associated with the chemotherapy, were able to reduce lipid peroxidation and protein oxidation, providing a beneficial immunological balance between the production of pro- and anti-inflammatory cytokines.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Supelco
Trimethoprim, VETRANAL®, analytical standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Diphenyl diselenide, 98%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%