Merck
Hepatic stellate cells induce immunotolerance of islet allografts.
Transplantation proceedings (2014-06-18)
Z-Y Zhang, Z-Q Zhou, K-B Song, S-C Kim, G-W Zhou
ABSTRAKT
Activated hepatic stellate cells (HSCs) possess strong immune inhibitory activity. The present study highlighted the protective role of HSCs in islet transplantation. Recipients were randomly divided into 4 groups: a diabetic group, an HSC-alone group, an islet-alone transplant group, and a cotransplant group. Graft survival was compared among the 4 groups. Serum transforming growth factor β (TGFβ), tumor necrosis factor α, interleukin-1β, and interferon gamma expression levels were measured. The infiltration of lymphocytes was observed via hematoxylin and eosin staining, and immunohistochemical examinations were performed. Results showed that allogeneic HSCs protect islet allografts better than syngeneic HSCs. There was significant prolonged graft survival and a higher level of TGFβ in the cotransplant group (P < .01). The infiltration of lymphocytes in the cotransplant group was notably less than in the islet-alone group (P < .01). The formation of desmin-positive HSC packages was detected in the cotransplant group. In conclusion, allogeneic HSCs can better prolong the survival of islet allografts by stimulating TGFβ expression and forming a biological capsule around the graft.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Dithizone, ACS reagent, for spectrophotometric det. of Cd, Cu, Hg, Pb, Zn, ≥98.0% (TLC)
Sigma-Aldrich
Dithizone, Practical Grade
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O