Merck
Degradation of amyloid beta by human induced pluripotent stem cell-derived macrophages expressing Neprilysin-2.
Stem cell research (2014-12-03)
Koutaro Takamatsu, Tokunori Ikeda, Miwa Haruta, Keiko Matsumura, Yasuhiro Ogi, Naomi Nakagata, Makoto Uchino, Yukio Ando, Yasuharu Nishimura, Satoru Senju
ABSTRAKT
The purpose of this study was to evaluate the therapeutic potential of human induced pluripotent stem (iPS) cell-derived macrophage-like cells for Alzheimer's disease (AD). In previous studies, we established the technology to generate macrophage-like myeloid lineage cells with proliferating capacity from human iPS cells, and we designated the cells iPS-ML. iPS-ML reduced the level of Aβ added into the culture medium, and the culture supernatant of iPS-ML alleviated the neurotoxicity of Aβ. We generated iPS-ML expressing the Fc-receptor-fused form of a single chain antibody specific to Aβ. In addition, we made iPS-ML expressing Neprilysin-2 (NEP2), which is a protease with Aβ-degrading activity. In vitro, expression of NEP2 but not anti-Aβ scFv enhanced the effect to reduce the level of soluble Aβ oligomer in the culture medium and to alleviate the neurotoxicity of Aβ. To analyze the effect of iPS-ML expressing NEP2 (iPS-ML/NEP2) in vivo, we intracerebrally administered the iPS-ML/NEP2 to 5XFAD mice, which is a mouse model of AD. We observed significant reduction in the level of Aβ in the brain interstitial fluid following administration of iPS-ML/NEP2. These results suggested that iPS-ML/NEP2 may be a potential therapeutic agent in the treatment of AD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Supelco
1,1,1,3,3,3-Hexafluoro-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)