Merck
  • Morphine and DAMGO produce an opposite effect on presynaptic glutamate release via different downstream pathways of μ opioid receptors in the basolateral amygdala.
Morphine and DAMGO produce an opposite effect on presynaptic glutamate release via different downstream pathways of μ opioid receptors in the basolateral amygdala.
Neuropharmacology (2014-09-10)
Jinhui Yang, Hualan Yang, Xiaowei Du, Qianqian Ma, Jiaojiao Song, Ming Chen, Yi Dong, Lan Ma, Ping Zheng
ABSTRAKT
Increasing evidence demonstrates that different opioids, while acting μ opioid receptors, can activate distinct downstream responses, a phenomenon termed functional selectivity or biased agonism. The present study designed experiments to test whether the μ receptor agonist morphine and D-Ala(2), N-Me-Phe(4), Gly(5)-ol-enkephalin (DAMGO) had a different effect on presynaptic glutamate release in the basolateral amygdala (BLA) and whether this difference was due to their biased agonism at μ receptors. The results showed that DAMGO markedly decreased the frequency of sEPSCs in pyramidal cells of BLA. The concentration-dependence experiment showed that DAMGO dose-dependently decreased the frequency of sEPSCs. Morphine markedly increased the frequency of sEPSCs in pyramidal cells of BLA. The concentration-dependence experiment showed that morphine dose-dependently increased the frequency of sEPSCs. We also used PPF of EPSC as another indicator of presynaptic glutamate release to confirm the opposite effect of morphine and DAMGO on the glutamate release. Further mechanism studies showed that the opposite effect of morphine and DAMGO on the glutamate release was via the activation of μ receptors, but the downstream signaling pathways of μ receptors were different: DAMGO inhibited the glutamate release via μ receptor-Gi protein- PLA2-AA signaling pathway, whereas morphine promoted the glutamate release via μ receptor-Gi protein-PKC-ERK1/2-synapsin I signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O