Merck
Downregulation of Sphingosine 1-Phosphate Receptor 1 Promotes the Switch from Tangential to Radial Migration in the OB.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-10-09)
Julieta Alfonso, Horst Penkert, Ceren Duman, Annalisa Zuccotti, Hannah Monyer
ABSTRAKT
Neuroblast migration is a highly orchestrated process that ensures the proper integration of newborn neurons into complex neuronal circuits. In the postnatal rodent brain, neuroblasts migrate long distances from the subependymal zone of the lateral ventricles to the olfactory bulb (OB) within the rostral migratory stream (RMS). They first migrate tangentially in close contact to each other and later radially as single cells until they reach their final destination in the OB. Sphingosine 1-phosphate (S1P) is a bioactive lipid that interacts with cell-surface receptors to exert different cellular responses. Although well studied in other systems and a target for the treatment of multiple sclerosis, little is known about S1P in the postnatal brain. Here, we report that the S1P receptor 1 (S1P1) is expressed in neuroblasts migrating in the RMS. Using in vivo and in vitro gain- and loss-of-function approaches in both wild-type and transgenic mice, we found that the activation of S1P1 by its natural ligand S1P, acting as a paracrine signal, contributes to maintain neuroblasts attached to each other while they migrate in chains within the RMS. Once in the OB, neuroblasts cease to express S1P1, which results in cell detachment and initiation of radial migration, likely via downregulation of NCAM1 and β1 integrin. Our results reveal a novel physiological function for S1P1 in the postnatal brain, directing the path followed by newborn neurons in the neurogenic niche. The function of each neuron is highly determined by the position it occupies within a neuronal circuit. Frequently, newborn neurons must travel long distances from their birthplace to their predetermined final location and, to do so, they use different modes of migration. In this study, we identify the sphingosine 1-phosphate (S1P) receptor 1 (S1P1) as one of the key players that govern the switch from tangential to radial migration of postnatally generated neuroblasts in the olfactory bulb. Of interest is the evidence that the ligand, S1P, is provided by nearby astrocytes. Finally, we also propose adhesion molecules that act downstream of S1P1 and initiate the transition from tangential chain migration to individual radial migration outside of the stream.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))