Merck
mGluR5 in the nucleus accumbens shell regulates morphine-associated contextual memory through reactive oxygen species signaling.
Addiction biology (2015-03-05)
Chong Qi, Xinjuan Wang, Feifei Ge, Yijing Li, Fang Shen, Junkai Wang, Cailian Cui
ABSTRAKT
Emerging evidence indicates that metabotropic glutamate receptor 5 (mGluR5) critically modulates drug and drug-related behaviors. However, the role of mGluR5 in the opiate-induced contextual memory remains unclear. Here, we found that microinfusion of the mGluR5 antagonist 3-((2-Methyl-1,3-thiazol-4-yl)ethynyl)pyridine (MTEP) into the nucleus accumbens (NAc) shell, but not into the core, significantly attenuated the expression of morphine conditioned place preference (CPP) in rats. Following the expression of morphine CPP, the protein level of membrane mGluR5 was selectively increased in the NAc shell. In primary striatal neurons, we observed that treatment with the mGluR5 agonist CHPG increased the phosphorylation level of extracellular signal-regulated kinase (ERK), which was dependent on the mGluR5-inositol-1,4,5-trisphosphate-reactive oxygen species (ROS) pathway. Moreover, the microinjection of the ROS scavenger Tempol into the NAc shell of rats blocked the expression of morphine CPP. Further, the administration of t-BOOH, a ROS donor, into the NAc shell rescued the retrieval impairment of morphine CPP produced by MTEP. Our previous study demonstrated that the expression of morphine CPP increased the phosphorylation of ERK selectively in the NAc shell. Thus, results of the present study suggest that mGluR5 in the NAc shell, but not in the core, is essential for the retrieval of morphine contextual memory, which is mediated at least in part, through the ROS/ERK signaling pathway. Uncovering the molecular basis of opiate contextual memory will benefit the development of new therapeutic approaches for the treatment of opiate addiction.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC