Merck
Removal of fluoxetine from water by adsorbent materials produced from paper mill sludge.
Journal of colloid and interface science (2015-02-25)
Guilaine Jaria, Vânia Calisto, María Victoria Gil, Marta Otero, Valdemar I Esteves
ABSTRAKT
In this work, three adsorbents were produced by using as precursor primary paper mill sludge and applying KOH, NaOH or ZnCl2 as chemical activating agents (PS800-10KOH, PS800-10NaOH and PS800-10ZnCl2) and subjected to pyrolysis. The produced materials were tested for removal of fluoxetine from water, reported as one of the most persistent pharmaceuticals in the aquatic environment. Also, primary sludge pyrolysed under the same conditions but without activation (PS800-10) and a commercial activated carbon (PBFG4) were studied for comparison. Physical and chemical properties of the materials were determined and adsorption kinetic and equilibrium studies were performed in batch experiments. Equilibrium studies allowed testing the capacity of the produced materials for adsorb fluoxetine-HCl, showing no apparent correlation between the S(BET) areas and the adsorption capacities of the materials. The maximum adsorption capacity (mg g(-1)) was of 191.6±4.8 for PS800-10KOH; 136.6±9.6 for PS800-10NaOH; 28.4±0.3 for PS800-10ZnCl2; 120.4±2.5 for PS800-10 and 96.2±1.0 for PBFG4. It was demonstrated that many factors influence the removal of fluoxetine from solution and that the surface area is not the main factor in the process. Also the activation process did not enhance the properties of the produced materials.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium tetraborate, 99%
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis