Merck
  • Clinical chemistry measurements with commercially available test slides on a smartphone platform: Colorimetric determination of glucose and urea.
Clinical chemistry measurements with commercially available test slides on a smartphone platform: Colorimetric determination of glucose and urea.
Clinica chimica acta; international journal of clinical chemistry (2015-06-24)
Yuanyuan Wu, Anukul Boonloed, Neal Sleszynski, Myra Koesdjojo, Chadd Armstrong, Shay Bracha, Vincent T Remcho
ABSTRAKT
Rapidly increasing healthcare costs in economically advantaged countries are currently unsustainable, while in many developing nations, even 50-year-old technologies are too expensive to implement. New and unconventional technologies are being explored as solutions to this problem. In this study, we examined the use of a smartphone as the detection platform for 2 well-developed, relatively inexpensive, commercially available clinical chemistry assays as a model for rapid and inexpensive clinical diagnostic testing. An Apple iPhone 4 camera phone equipped with a color analysis application (ColorAssist) was combined with Vitros® glucose and urea colorimetric assays. Color images of assay slides at various concentrations of glucose or urea were collected with the iPhone 4 and quantitated in three different spectral ranges (red/green/blue or RGB) using the ColorAssist app. When the diffuse reflectance data was converted into absorbance, it was possible to quantitate glucose or blood urea nitrogen (BUN) over their clinically important concentration ranges (30-515mg/dl for glucose or 2-190mg/dl for BUN), with good linearity (R(2)=0.9994 or 0.9996, respectively [n=5]). Data collected using the iPhone 4 and canine serum samples were in agreement with results from the instrumental "gold standard" (Beckman Coulter AU480 Chemistry System) (R(2)=0.9966 and slope=1.0001 for glucose; R(2)=0.9958 and slope=0.9454 for BUN). Glucose determinations of serum samples made using this smartphone method were as accurate as or more accurate than a commercial colorimetric dry slide analyzer (Heska® Element DC Chemistry Analyzer, Loveland, CO) and 2 glucometers: ReliOn® Ultima (Abbott Diabetes Care Inc) and Presto® (AgaMatrix Inc.H). BUN determinations made using the smartphone approach were comparable in accuracy to the Heska instrument. This demonstration shows that smartphones have the potential to be used as simple, effective colorimetric detectors for quantitative diagnostic tests, and may be applicable for both point-of-care applications in the developed world and field deployment in developing nations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)