Merck
Late Sodium Current in Human Atrial Cardiomyocytes from Patients in Sinus Rhythm and Atrial Fibrillation.
PloS one (2015-06-30)
Claire Poulet, Erich Wettwer, Morten Grunnet, Thomas Jespersen, Larissa Fabritz, Klaus Matschke, Michael Knaut, Ursula Ravens
ABSTRAKT
Slowly inactivating Na+ channels conducting "late" Na+ current (INa,late) contribute to ventricular arrhythmogenesis under pathological conditions. INa,late was also reported to play a role in chronic atrial fibrillation (AF). The objective of this study was to investigate INa,late in human right atrial cardiomyocytes as a putative drug target for treatment of AF. To activate Na+ channels, cardiomyocytes from transgenic mice which exhibit INa,late (ΔKPQ), and right atrial cardiomyocytes from patients in sinus rhythm (SR) and AF were voltage clamped at room temperature by 250-ms long test pulses to -30 mV from a holding potential of -80 mV with a 100-ms pre-pulse to -110 mV (protocol I). INa,late at -30 mV was not discernible as deviation from the extrapolated straight line IV-curve between -110 mV and -80 mV in human atrial cells. Therefore, tetrodotoxin (TTX, 10 μM) was used to define persistent inward current after 250 ms at -30 mV as INa,late. TTX-sensitive current was 0.27±0.06 pA/pF in ventricular cardiomyocytes from ΔKPQ mice, and amounted to 0.04±0.01 pA/pF and 0.09±0.02 pA/pF in SR and AF human atrial cardiomyocytes, respectively. With protocol II (holding potential -120 mV, pre-pulse to -80 mV) TTX-sensitive INa,late was always larger than with protocol I. Ranolazine (30 μM) reduced INa,late by 0.02±0.02 pA/pF in SR and 0.09±0.02 pA/pF in AF cells. At physiological temperature (37°C), however, INa,late became insignificant. Plateau phase and upstroke velocity of action potentials (APs) recorded with sharp microelectrodes in intact human trabeculae were more sensitive to ranolazine in AF than in SR preparations. Sodium channel subunits expression measured with qPCR was high for SCN5A with no difference between SR and AF. Expression of SCN8A and SCN10A was low in general, and lower in AF than in SR. In conclusion, We confirm for the first time a TTX-sensitive current (INa,late) in right atrial cardiomyocytes from SR and AF patients at room temperature, but not at physiological temperature. While our study provides evidence for the presence of INa,late in human atria, the potential of such current as a target for the treatment of AF remains to be demonstrated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Cesium methanesulfonate, used in patch clamp techniques