Merck
Analysis of immune cell populations and cytokine profiles in murine splenocytes exposed to whole-body low-dose irradiation.
International journal of radiation biology (2015-07-03)
Kyung-Hee Song, Mi-Hyoung Kim, Seong-Mook Kang, Seung-Youn Jung, Jiyeon Ahn, Hee-Jong Woo, Seon Young Nam, Sang-Gu Hwang, Sang-Young Ryu, Jie-Young Song
ABSTRAKT
In contrast to high-dose therapeutic irradiation, definitive research detailing the physiological effects of low-dose irradiation is limited. Notably, the immunological response elicited after low-dose irradiation remains controversial. Female C57BL/6 mice were whole- body-irradiated with a single or three daily fractions up to a total dose of 0.1, 1, or 10 cGy. Blood and spleen were harvested 2, 7 and 14 days after irradiation. The splenic CD4(+) T cell subpopulations were temporarily increased at 2 days after single or fractionated irradiation, whereas the percentage of dendritic cells (DC) and macrophages was decreased. Whereas CD8(+) T cell populations were decreased in single-dose irradiated mice at day 7, early and sustained reduction of CD8(+) T cell numbers was observed in fractionated- dose-irradiated mice from day 2 until day 14. In addition, single-dose irradiation resulted in a Th1 cytokine expression profile, whereas fractionated-dose irradiation drove a Th2 shift. Additionally, increased expression of immune-related factors was observed at early time-points with single-dose irradiation, in contrast to the dose-independent induction following fractionated-dose irradiation. Our results demonstrate that low-dose irradiation modulates the immune response in mice, where the sensitivity and kinetics of the induced response vary according to the dosing method.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Potassium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%