Merck
The Role of D4Z4-Encoded Proteins in the Osteogenic Differentiation of Mesenchymal Stromal Cells Isolated from Bone Marrow.
Stem cells and development (2015-07-21)
Laurence de la Kethulle de Ryhove, Eugénie Ansseau, Charlotte Nachtegael, Karlien Pieters, Céline Vanderplanck, Mieke Geens, Karen Sermon, Steve D Wilton, Frédérique Coppée, Laurence Lagneaux, Alexandra Belayew
ABSTRAKT
Facioscapulohumeral muscular dystrophy (FSHD) is associated with an activation of the double homeobox 4 (DUX4) gene, which we previously identified within the D4Z4 repeated elements in the 4q35 subtelomeric region. The pathological DUX4 mRNA is derived from the most distal D4Z4 unit and extends unexpectedly within the flanking pLAM region, which provides an intron and polyadenylation signal. The conditions that are required to develop FSHD are a permissive allele providing the polyadenylation signal and hypomethylation of the D4Z4 repeat array compared with the healthy muscle. The DUX4 protein is a 52-kDa transcription factor that initiates a large gene deregulation cascade leading to muscle atrophy, inflammation, differentiation defects, and oxidative stress, which are the key features of FSHD. DUX4 is a retrogene that is normally expressed in germline cells and is submitted to repeat-induced silencing in adult tissues. Since DUX4 mRNAs have been detected in human embryonic and induced pluripotent stem cells, we investigated whether they could also be expressed in human mesenchymal stromal cells (hMSCs). We found that DUX4 mRNAs were induced during the differentiation of hMSCs into osteoblasts and that this process involved DUX4 and new longer protein forms (58 and 70 kDa). A DUX4 mRNA with a more distant 5' start site was characterized that presented a 60-codon reading frame extension and encoded the 58-kDa protein. Transfections of hMSCs with an antisense oligonucleotide targeting DUX4 mRNAs decreased both the 52- and 58-kDa protein levels and confirmed their identity. Gain- and loss-of-function experiments in hMSCs suggested these DUX4 proteins had opposite roles in osteogenic differentiation as evidenced by the alkaline phosphatase activity and calcium deposition. Differentiation was delayed by the 58-kDa DUX4 expression and it was increased by 52-kDa DUX4. These data indicate a role for DUX4 protein forms in the osteogenic differentiation of hMSCs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%