Merck
Crystallographic analysis of the Arabidopsis thaliana BAG5-calmodulin protein complex.
Acta crystallographica. Section F, Structural biology communications (2015-07-07)
Boyang Cui, Shasha Fang, Yangfei Xing, Yuequan Shen, Xue Yang
ABSTRAKT
Arabidopsis thaliana BAG5 (AtBAG5) belongs to the plant BAG (Bcl-2-associated athanogene) family that performs diverse functions ranging from growth and development to abiotic stress and senescence. BAG family members can act as nucleotide-exchange factors for heat-shock protein 70 (Hsp70) through binding of their evolutionarily conserved BAG domains to the Hsp70 ATPase domain, and thus may be involved in the regulation of chaperone-mediated protein folding in plants. AtBAG5 is distinguished from other family members by the presence of a unique IQ motif adjacent to the BAG domain; this motif is specific for calmodulin (CaM) binding, indicating a potential role in the plant calcium signalling pathway. To provide a better understanding of the IQ motif-mediated interaction between AtBAG5 and CaM, the two proteins were expressed and purified separately and then co-crystallized together. Diffraction-quality crystals of the complex were grown using the sitting-drop vapour-diffusion technique from a condition consisting of 0.1 M Tris-HCl pH 8.5, 2.5 M ammonium sulfate. The crystals belonged to space group P2(1)2(1)2(1), with unit-cell parameters a = 64.56, b = 74.89, c = 117.09 Å. X-ray diffraction data were recorded to a resolution of 2.5 Å from a single crystal using synchrotron radiation. Assuming the presence of two molecules in the asymmetric unit, a Matthews coefficient of 2.44 Å(3) Da(-1) was calculated, corresponding to a solvent content of approximately 50%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture