Merck
  • Lubricin/Proteoglycan 4 Binding to CD44 Receptor: A Mechanism of the Suppression of Proinflammatory Cytokine-Induced Synoviocyte Proliferation by Lubricin.
Lubricin/Proteoglycan 4 Binding to CD44 Receptor: A Mechanism of the Suppression of Proinflammatory Cytokine-Induced Synoviocyte Proliferation by Lubricin.
Arthritis & rheumatology (Hoboken, N.J.) (2015-02-25)
Afnan Al-Sharif, Maha Jamal, Ling X Zhang, Katherine Larson, Tannin A Schmidt, Gregory D Jay, Khaled A Elsaid
ABSTRAKT
To evaluate the binding of recombinant human proteoglycan 4 (rhPRG4) to CD44 receptor and its consequences on cytokine-induced synoviocyte proliferation. The binding of rhPRG4 to CD44 and competition with high molecular weight (HMW) hyaluronic acid (HA) was evaluated using a direct enzyme-linked immunosorbent assay (ELISA) and surface plasmon resonance. Sialidase A and O-glycosidase digestion of rhPRG4 was performed, and CD44 binding was evaluated using ELISA. Rheumatoid arthritis (RA) fibroblast-like synoviocytes (FLS) were stimulated with interleukin-1β (IL-1β) or tumor necrosis factor α (TNFα) for 48 hours in the presence or absence of rhPRG4 or HMW HA at 20, 40, and 80 μg/ml, and cell proliferation was measured. The contribution of CD44 was assessed by coincubation with a CD44 antibody (IM7). The antiproliferative effect of rhPRG4 was investigated following treatment of PRG4(-/-) mouse synoviocytes with IL-1β or TNFα in the presence or absence of IM7. Recombinant human PRG4 bound CD44 and interfered with the binding of HMW HA to CD44. Removal of sialic acid and O-glycosylations significantly increased CD44 binding by rhPRG4 (P < 0.001). Both rhPRG4 and HMW HA at 40 and 80 μg/ml significantly suppressed IL-1β-induced proliferation of RA FLS (P < 0.05). Recombinant human PRG4 at 20, 40, and 80 μg/ml significantly suppressed TNFα-induced RA FLS proliferation (P < 0.05). CD44 neutralization reversed the effect of rhPRG4 on IL-1β- and TNFα-stimulated RA FLS and the effect of HMW HA on IL-1β-stimulated RA FLS. Recombinant human PRG4 inhibited cytokine-induced proliferation of PRG4(-/-) synoviocytes, which could be prevented by blocking CD44. PRG4 (lubricin) is a novel putative ligand for CD44 and may control synoviocyte overgrowth in inflammatory arthropathies via a CD44-mediated mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
Sodium chloride solution, 5 M