Merck
Proteomic analysis of cell wall in four pathogenic species of Candida exposed to oxidative stress.
Microbial pathogenesis (2015-07-19)
Mayra Denisse Ramírez-Quijas, Everardo López-Romero, Mayra Cuéllar-Cruz
ABSTRAKT
In order for Candida species to adhere and colonize human host cells they must express cell wall proteins (CWP) and adapt to reactive oxygen species (ROS) generated by phagocytic cells of the human host during the respiratory burst. However, how these pathogens change the expression of CWP in response to oxidative stress (OSR) is not known. Here, fifteen moonlight-like CWP were identified that expressed differentially in four species of Candida after they were exposed to H2O2 or menadione (O2(-)). These proteins included: (i) glycolytic enzymes, such as glyceraldehyde-3-phosphate dehydrogenase (Gapdh), fructose-bisphosphate aldolase (Fba1), phosphoglycerate mutase (Gpm1), phosphoglycerate kinase (Pgk), pyruvate kinase (Pk) and enolase (Eno1); (ii) the heat shock proteins Ssb1 and Ssa2; (iii) OSR proteins such as peroxyredoxin (Tsa1), the stress protein Ddr48 (Ddr48) and glutathione reductase (Glr1); (iv) other metabolic enzymes such as ketol-acid reductoisomerase (Ilv5) and pyruvate decarboxylase (Pdc1); and (v) other proteins such as elongation factor 1-beta (Efb1) and the 14-3-3 protein homolog. RT-PCR revealed that transcription of the genes coding for some of the identified CWP are differentially regulated. To our knowledge this is the first report showing that moonlight-like CWP are the first line of defense of Candida against ROS, and that they are differentially regulated in each of these pathogens.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg