Merck
Potato Virus M-Like Nanoparticles: Construction and Characterization.
Molecular biotechnology (2015-09-09)
Ieva Kalnciema, Ina Balke, Dace Skrastina, Velta Ose, Andris Zeltins
ABSTRAKT
Virus-like particles (VLPs) are multisubunit self-assembly competent protein structures with identical or highly related overall structure to their corresponding native viruses. To construct a new filamentous VLP carrier, the coat protein (CP) gene from potato virus M (PVM) was amplified from infected potato plants, cloned, and expressed in Escherichia coli cells. As demonstrated by electron microscopy analysis, the PVM CP self-assembles into filamentous PVM-like particles, which are mostly 100-300 nm in length. Adding short Gly-Ser peptide at the C-terminus of the PVM, CP formed short VLPs, whereas peptide and protein A Z-domain fusions at the CP N-terminus retained its ability to form typical PVM VLPs. The PVM-derived VLP carrier accommodates up to 78 amino acid-long foreign sequences on its surface and can be produced in technologically significant amounts. PVM-like particles are stable at physiological conditions and also, apparently do not become disassembled in high salt and high pH solutions as well as in the presence of EDTA or reducing agents. Despite partial proteolytic processing of doubled Z-domain fused to PVM VLPs, the rabbit IgGs specifically bind to the particles, which demonstrates the functional activity and surface location of the Z-domain in the PVM VLP structure. Therefore, PVM VLPs may be recognized as powerful structural blocks for new human-made nanomaterials.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Kwas 3-hydroksypirydyno-2-karboksylowy, 98%