MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Oligodontia and curly hair occur with ectodysplasin-a mutations.
Journal of dental research (2014-02-04)
K E Lee, J Ko, T J Shin, H K Hyun, S H Lee, J W Kim
ABSTRACT
Oligodontia is the developmental absence of more than 5 permanent teeth except for the third molar. Familial oligodontia can occur as an isolated form or as part of a genetic syndrome. Mutations in the MSX1, PAX9, AXIN2, EDA, and WNT10A genes have been identified in familial non-syndromic oligodontia. Ectodermal dysplasia is a group of syndromes involving abnormalities of the ectodermal structures and is comprised of more than 150 different forms. Mutations in the ectodysplasin-A (EDA) gene have been associated with X-linked hypohidrotic ectodermal dysplasia, and partial disruption of the EDA signaling pathway has been shown to cause an isolated form of oligodontia. We identified 2 X-linked oligodontia families and performed mutational analysis of the EDA gene. The mutational analysis revealed 2 novel EDA mutations: c.866G>T, p.Arg289Leu and c.1135T>G, p.Phe379Val (reference sequence NM_001399.4). These mutations were perfectly segregated with oligodontia and curly hair within each family and were not found in the 150 control X-chromosomes with the same ethnic background and in the exome variant server. This study broadens the mutational spectrum of the EDA gene and the understanding of X-linked oligodontia with curly hair.
Sigma-Aldrich
L-Leucine, reagent grade, ≥98% (HPLC)
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Valine
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucine, 99%, FG
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Guanine, BioUltra
Sigma-Aldrich
Thymine, ≥99%
Sigma-Aldrich
Thymine, BioReagent, suitable for cell culture
Supelco
Thymine(Zidovudine Related Compound C), Pharmaceutical Secondary Standard; Certified Reference Material
Leucine, European Pharmacopoeia (EP) Reference Standard
Valine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Phenylalanine, SAJ special grade, ≥99.0%
SAFC
L-Arginine
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Arginine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Phenylalanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%