MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Gastrointestinal effects of the addition of ascorbic acid to aspirin.
Pain practice : the official journal of World Institute of Pain (2011-12-14)
Vrunda Patel, Matt Fisher, Michael Voelker, Uwe Gessner
ABSTRACT
Nonsteroidal anti-inflammatory drugs (NSAIDs), including aspirin, have been associated with the damage to the gastrointestinal tract. One proposed mechanism of injury to the gastrointestinal mucosa by NSAIDs is oxygen radical-dependent microvascular injury. There is reasonable evidence to support the benefit of the addition of ascorbic acid, an ingredient with antioxidant properties, to moderate the adverse gastrointestinal (GI) effects of aspirin. Pharmacokinetic data have demonstrated that aspirin and ascorbic acid combination therapy can assist in mitigating the decrease in levels of ascorbic acid secondary to aspirin monotherapy. Endoscopic evaluation has demonstrated that the addition of ascorbic acid to aspirin significantly improves Lanza scores and rates of blood loss when compared to aspirin administration alone. When taken with ascorbic acid, the patient-reported tolerability of aspirin has been shown to be comparable to paracetamol and placebo. The existing body of evidence is relevant to short-term therapy with analgesic aspirin doses, and extrapolation to long-term therapy with low-dose aspirin is not appropriate. The purported benefit of an aspirin and ascorbic acid combination is a local observance and is not suspected to influence the adverse GI effects experienced as a result of systemic prostaglandin inhibition. Nevertheless, ascorbic acid may be a viable addition to the strategies employed to improve the gastrointestinal tolerability of aspirin.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline