MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • RFX-1-dependent activation of SHP-1 inhibits STAT3 signaling in hepatocellular carcinoma cells.
RFX-1-dependent activation of SHP-1 inhibits STAT3 signaling in hepatocellular carcinoma cells.
Carcinogenesis (2014-10-18)
Jung-Chen Su, Heng-Chieh Chiang, Ping-Hui Tseng, Wei-Tien Tai, Cheng-Yi Hsu, Yong-Shi Li, Jui-Wen Huang, Ching-Huai Ko, Mai-Wei Lin, Pei-Yi Chu, Chun-Yu Liu, Kuen-Feng Chen, Chung-Wai Shiau
ABSTRACT
Regulatory factor X-1 (RFX-1) is a transcription factor that has been linked to negative regulation of tumor progression; however, its biological function and signaling cascades are unknown. Here, we performed several studies to elucidate the roles of RFX-1 in the regulation of SHP-1 in hepatocellular carcinoma (HCC) cells. Overexpression of RFX-1 resulted in the activation of SHP-1 and repressed colony formation of HCC cells. In addition, by a mouse xenograft model, we demonstrated that RFX-1 overexpression also inhibited the tumor growth of HCC cells in vivo, suggesting that RFX-1 is of potential interest for small-molecule-targeted therapy. We also found that SC-2001, a bipyrrole molecule, induced apoptosis in HCC cells through activating RFX-1 expression. SC-2001 induced RFX-1 translocation from the cytosol to nucleus, bound to the SHP-1 promoter, and activated SHP-1 transcription. In a xenograft model, knockdown of RFX-1 reversed the antitumor effect of SC-2001. Notably, SC-2001 is much more potent than sorafenib, a clinically approved drug for HCC, in in vitro and in vivo assays. Our study confirmed that RFX-1 acts as a tumor suppressor in HCC and might be a new target for HCC therapy. The findings of this study also provide a new lead compound for targeted therapy via the activation of the RFX-1/SHP-1 pathway.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)