MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Optimal extraction and fingerprinting of carotenoids by accelerated solvent extraction and liquid chromatography with tandem mass spectrometry.
Optimal extraction and fingerprinting of carotenoids by accelerated solvent extraction and liquid chromatography with tandem mass spectrometry.
Food chemistry (2015-02-11)
Supradip Saha, Suresh Walia, Aditi Kundu, Khushbu Sharma, Ranjit Kumar Paul
ABSTRACT
Accelerated solvent extraction (ASE) is applied for the extraction of carotenoids from orange carrot and the extraction parameters were optimized. Two carotenoids, lutein and β-carotene, are selected as the validation process. Hildebrand solubility parameters and dielectric constant of solvents were taken into consideration in selecting solvent mixture. The effects of various experimental parameters, such as temperature, static time, drying agent etc., on the ASE extraction efficiency are investigated systematically. Interactions among the variables were also studied. Furthermore, two carotenoids were analyzed and characterized by LC-ESI MS. The study concluded that Hildebrand solubility parameter approach may be applicable for less polar bioactive molecules like carotenoids. The properties of solvent and extraction temperature are found to be the most important parameters affecting the ASE extraction efficiency of thermolabile natural compounds.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Xanthophyll, from marigold
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%