MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Development and validation of liquid chromatography-tandem mass spectrometry method for simultaneous determination of six steroidal saponins in rat plasma and its application to a pharmacokinetics study.
Development and validation of liquid chromatography-tandem mass spectrometry method for simultaneous determination of six steroidal saponins in rat plasma and its application to a pharmacokinetics study.
Steroids (2015-01-27)
Zhirui Liu, Wenxing Qin, Zhenyu Zhu, Yao Liu, Fengjun Sun, Yifeng Chai, Peiyuan Xia
ABSTRACT
A specific and reliable liquid chromatography-electrospray ionization-tandem mass spectrometry method was developed for the simultaneous determination of timosaponin H1 (TH1), timosaponin E1 (TE1), timosaponin E (TE), timosaponin B-II (TB-II), timosaponin B-III (TB-III) and anemarrhenasaponin I (AS-I) in rat plasma. After addition of internal standard (IS) ginsenoside Rh1, plasma samples were pretreated by protein precipitation with acetonitrile. Chromatographic separation was performed on a reverse phase ACQUITY™ BEH C18 column (100mm×2.1mm i.d., 1.7μm) using a gradient mobile phase system of acetonitrile-water containing 0.05% formic acid and 5mM ammonium formate. The triple quadruple mass spectrometer was set in negative electrospray ionization mode and multiple reaction monitoring (MRM) was used for six steroidal saponins quantification. The precursors to produce ion transitions monitored for TH1, TE1, TE, TB-II, TB-III, AS-I and IS were m/z 1211.5>1079.6, 935.5>773.4, 935.4>773.5, 919.6>757.4, 901.5>739.3, 757.4>595.3 and 637.3>475.3, respectively. The method validation was conducted over the curve range of 0.5-400ng/mL for the six saponins. The intra- and inter-day precisions (RSD%) were less than 9.4% and the average extraction recoveries ranged from 82.5% to 97.8% for each analyte. Six steroidal saponins were proved to be stable during sample storage, preparation and analytical procedures. The validated method was successfully applied for the first time to determine the concentrations of six main steroidal saponins in incurred rat plasma samples, after intragastric administration of the extract of Anemarrhena asphodeloides Bge. for a rat pharmacokinetic study.
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography