MilliporeSigma
  • The effect of stable macromolecular complexes of ionic polyphosphazene on HIV Gag antigen and on activation of human dendritic cells and presentation to T-cells.
The effect of stable macromolecular complexes of ionic polyphosphazene on HIV Gag antigen and on activation of human dendritic cells and presentation to T-cells.
Biomaterials (2014-07-16)
Christine D Palmer, Jana Ninković, Zofia M Prokopowicz, Christy J Mancuso, Alexander Marin, Alexander K Andrianov, David J Dowling, Ofer Levy
ABSTRACT
Neonates and infants are susceptible to infection due to distinct immune responses in early life. Therefore, development of vaccine formulation and delivery systems capable of activating human newborn leukocytes is of global health importance. Poly[di(carboxylatophenoxy)phosphazene] (PCPP) belongs to a family of ionic synthetic polyphosphazene polyelectrolyte compounds that can form non-covalent interactions with protein antigens and demonstrate adjuvant activity in animals and in human clinical trials. However, little is known about their ability to activate human immune cells. In this study, we characterized the effects of PCPP alone or in combination with a model antigen (recombinant HIV-Gag (Gag)), on the maturation, activation and antigen presentation by human adult and newborn dendritic cells (DCs) in vitro. PCPP treatment induced DC activation as assessed by upregulation of co-stimulatory molecules and cytokine production. Studies benchmarking PCPP to Alum, the most commonly used vaccine adjuvant, demonstrated that both triggered cell death and release of danger signals in adult and newborn DCs. When complexed with Gag antigen, PCPP maintained its immunostimulatory characteristics while permitting internalization and presentation of Gag by DCs to HIV-Gag-specific CD4(+) T cell clones. The PCPP vaccine formulation outlined here has intrinsic adjuvant activity, can facilitate effective delivery of antigen to DCs, and may be advantageous for induction of beneficial T cell-mediated immunity. Moreover, polyphosphazenes can further reduce cost of vaccine production and distribution through their dose-sparing and antigen-stabilizing properties, thus potentially eliminating the need for cold chain distribution.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture