MilliporeSigma
  • Protein malnutrition alters spleen cell proliferation and IL-2 and IL-10 production by affecting the STAT-1 and STAT-3 balance.
Protein malnutrition alters spleen cell proliferation and IL-2 and IL-10 production by affecting the STAT-1 and STAT-3 balance.
Inflammation (2014-07-06)
Alexandra Siqueira Mello, Dalila Cunha de Oliveira, Bruna Bizzarro, Anderson Sá-Nunes, Araceli Aparecida Hastreiter, Jackeline Soares de Oliveira Beltran, José Guilherme Xavier, Primavera Borelli, Ricardo Ambrósio Fock
ABSTRACT
Protein malnutrition (PM) is an important public health problem that affects resistance to infection by impairing a number of physiological processes. PM induces structural changes in the lymphoid organs that affect the roles of the immune and inflammatory responses in a crucial way. The activation of different transcription factors, including signal transducer and activator of transcription (STAT) family members, leads to the production of different cytokines, which are mediators essential to mounting adequate immune and inflammatory responses. In this study, malnourished animals presented anemia, leukopenia, and a severe reduction in spleen cellularity, with reduced numbers of most cell populations, as well as increased percentages of CD3(+) and CD4(+) cells. The proliferation rates were reduced, and cells were increasingly observed in the G0/G1 cell cycle phase; further, IL-2 production was reduced, while IL-10 production was increased. In spleen cells from malnourished animals, STAT-3 protein expression was increased, with a concomitant reduction in STAT-1 expression. Knowing that STAT-1 and STAT-3 are key transcription factors in both immunity and inflammatory pathways, these results infer, at least in part, a mechanistic pathway that affects the manner or intensity of the immune response in malnourished individuals, increasing susceptibility to infection.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), Type IV-S, lyophilized powder, aseptically processed, BioReagent, suitable for cell culture