MilliporeSigma
  • Fabrication of monodisperse liposomes-in-microgel hybrid microparticles in capillary-based microfluidic devices.
Fabrication of monodisperse liposomes-in-microgel hybrid microparticles in capillary-based microfluidic devices.
Colloids and surfaces. B, Biointerfaces (2014-10-08)
Eun Seon Jeong, Han Am Son, Min Kyung Kim, Kyoung-Ho Park, Sechan Kay, Pil Seok Chae, Jin Woong Kim
ABSTRACT
This study introduces a drop-based microfluidic approach to physically immobilize liposomes in microgel (liposomes-in-microgel) particles. For this, we generate a uniform liposomes-in-water-in-oil emulsion in a capillary-based microfluidic device. Basically, we have investigated how the flow rate and flow composition affect generation of emulsion precursor drops in micro-channels. Then, the precursor emulsion drops are solidified by photo-polymerization. From characterization of hydrogel mesh sizes, we have figured out that the mesh size of the liposomes-in-microgel particles is bigger than that of bare microgel particles, since liposomes take space in the hydrogel phase. In our further study on drug releasing, we have observed that immobilization of liposomes in the microgel particles can not only remarkably retard drug releasing, but also enables a sustained release, which stems from the enhanced matrix viscosity of the surrounding hydrogel phase.
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), ≥99.0%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard