MilliporeSigma
  • Functional expression, characterization and application of the human S100A4 protein.
Functional expression, characterization and application of the human S100A4 protein.
Molecular medicine reports (2014-10-24)
Degang Wang, Jianwei Zhang, Ziquan Liu, Yunyun Chen, Chuanxiang Xu, Zhiqing Zhang, Xiaohua Liu, Lei Wu, Xuesi Zhou, Xiangyan Meng, Hua Li, Hongtao Liu, Zifeng Jiang, Tianhui Wang
ABSTRACT
Preparations utilizing monoclonal antibodies against S100A4 provide useful tools for functional studies to investigate the clinical applications of the human S100A4 protein. In the present study, human S100A4 protein was expressed in Escherichia coli (E. coli) BL21 (DE3), successfully purified by diethylaminoethyl cellulose anion-exchange chromatography and identified by western blot analysis. Soluble S100A4 bioactivity was confirmed by Transwell migration and invasion assays in the human HeLa cell line. Monoclonal antibodies (mAbs) were generated utilizing the standard hybridoma method and were validated by enzyme-linked immunosorbent assay and western blot analysis. The antibody was then used to examine human gastric carcinoma specimens by immunohistochemistry. Recombinant S100A4 was functionally expressed in E. coli and promoted the migration and invasion of HeLa cells. Four hybridoma cell lines, which secreted mAbs specifically against human S100A4 protein, were obtained. One of the four mAbs, namely 2A12D10B2, recognized human S100A4 as indicated by immunohistochemical staining of human gastric carcinoma specimens and recombinant S100A4 was functionally expressed in E. coli. The mAbs of recombinant S100A4 were suitable for detecting S100A4 expression in human tissues and for investigating the subsequent clinical applications of the protein.
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), ≥99.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet