MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Lymphocyte inhibition is compromised in mesenchymal stem cells from psoriatic skin.
Lymphocyte inhibition is compromised in mesenchymal stem cells from psoriatic skin.
European journal of dermatology : EJD (2014-12-03)
Ruifeng Liu, Yong Wang, Xincheng Zhao, Yuanwen Yang, Kaiming Zhang
ABSTRACT
Psoriasis is a chronic inflammatory skin disorder associated with a host of immune abnormalities. Mesenchymal stem cells (MSCs) have immunosuppressive properties and, in earlier studies, we found that the bone marrow MSCs of patients with psoriasis exhibit abnormal cytokine secretion. Since MSCs can be isolated from skin, we hypothesized that the biological characteristics of MSCs in psoriatic skin lesions might reflect the pathogenesis of psoriasis. To investigate the effects of MSCs from psoriatic skin lesions on T-cell proliferation. MSCs obtained from psoriatic skin lesions and healthy human skin were examined by flow cytometry and cell differentiation assays. MSCs were co-cultured with normal peripheral blood T cells to assess changes in T-cell proliferation. Concentrations of interleukin (IL)-6, IL-11, hepatocyte growth factor (HGF), and transforming growth factor (TGF)-β1 in the MSC culture supernatants were measured by enzyme-linked immunosorbent assays. Surface markers and differentiation capacity were similar in MSCs from both sources. MSCs in psoriatic skin lesions were weaker inhibitors of T-cell proliferation (p<0.05) and exhibited increased secretion of IL-11 and reduced secretion of IL-6 and HGF (p<0.05). Secretion of TGF-β1 was unchanged (p > 0.05). This study demonstrated abnormalities in MSCs derived from psoriatic skin lesions. We suggest that the attenuated inhibitory effect on T-cell proliferation might be one of the pathogenic mechanisms of psoriasis.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture