MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Prion protein and copper cooperatively protect neurons by modulating NMDA receptor through S-nitrosylation.
Prion protein and copper cooperatively protect neurons by modulating NMDA receptor through S-nitrosylation.
Antioxidants & redox signaling (2014-12-10)
Lisa Gasperini, Elisa Meneghetti, Beatrice Pastore, Federico Benetti, Giuseppe Legname
ABSTRACT
Several neurodegenerative disorders show alterations in glutamatergic synapses and increased susceptibility to excitotoxicity. Mounting evidence suggests a central role for the cellular prion protein (PrP(C)) in neuroprotection. Therefore, the loss of PrP(C) function occurring in prion disorders may contribute to the disease progression and neurodegeneration. Indeed, PrP(C) modulates N-methyl-d-aspartate receptors (NMDAR), thus preventing cell death. In this study, we show that PrP(C) and copper cooperatively inhibit NMDAR through S-nitrosylation, a post-translational modification resulting from the chemical reaction of nitric oxide (NO) with cysteines. Comparing wild-type Prnp (Prnp(+/+)) and PrP(C) knockout (Prnp(0/0)) mouse hippocampi, we found that GluN1 and GluN2A S-nitrosylation decrease in Prnp(0/0). Using organotypic hippocampal cultures, we found that copper chelation decreases NMDAR S-nitrosylation in Prnp(+/+) but not in Prnp(0/0). This suggests that PrP(C) requires copper to support the chemical reaction between NO and thiols. We explored PrP(C)-Cu neuroprotective role by evaluating neuron susceptibility to excitotoxicity in Prnp(+/+) and Prnp(0/0) cultures. We found that (i) PrP(C)-Cu modulates GluN2A-containing NMDAR, those inhibited by S-nitrosylation; (ii) PrP(C) and copper are interdependent to protect neurons from insults; (iii) neuronal NO synthase inhibition affects susceptibility in wild-type but not in Prnp(0/0), while (iv) the addition of a NO donor enhances Prnp(0/0) neurons survival. Our results show that PrP(C) and copper support NMDAR S-nitrosylation and cooperatively exert neuroprotection. In addition to NMDAR, PrP(C) may also favor the S-nitrosylation of other proteins. Therefore, this mechanism may be investigated in the context of the different cellular processes in which PrP(C) is involved.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist
Sigma-Aldrich
Plasma, from chicken
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
S-Methyl methanethiosulfonate, purum, ≥98.0% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
S-Methyl methanethiosulfonate, 97%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)