MilliporeSigma
  • Tandem expression in E. coli of type III PKS and P450 genes from marine Streptomyces olivaceus FXJ 7.023 gives production of phenol and indole.
Tandem expression in E. coli of type III PKS and P450 genes from marine Streptomyces olivaceus FXJ 7.023 gives production of phenol and indole.
World journal of microbiology & biotechnology (2015-02-24)
Changwu Yue, Ning Liu, Minghao Liu, Yuhong Lü, Meiyun Shao, Miao Wang, Guoming Ai, Ying Huang
ABSTRACT
The draft genome sequence of marine Streptomyces olivaceus strain FXJ 7.023 contains a cryptic Type III polyketide synthase (type III PKS) gene cluster, which is similar to the Streptomyces coelicolor THN biosynthesis gene cluster. A putative type III PKS (SoRppA) gene and its adjacent gene for cytochrome P450 158A2 (SoCYP158A2) of this gene cluster were cloned by PCR screening through a fosmid genomic library of S. olivaceus FXJ 7.023. Tandem expression of SoRppA and SoCYP158A2 in Escherichia coli strain BL21 (DE3) plysS resulted in obvious biosynthesis of phenol and indole, while heterologous expression of SoRppA or SoCYP158A2 alone did not. The engineered strain sorppAcyp158a2BL21 showed tolerance to phenol concentration up to 0.75 g/L. Continuous biosynthesis of phenol and indole by the immobilized engineered strain on macroporousresin was achieved, and the productivities of phenol and indole in extractant-free culture in 102 h were 0.08 and 1.525 g/L/h, respectively, with the highest production reached 0.67 and 14.48 g/L, respectively. These results suggest that the engineered strain and immobilized continuous fermentation process may provide potential for "green" production of phenol and indole.
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Nitrogen, ≥99.998%
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard