MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Identification of polyphenols from antiviral Chamaecrista nictitans extract using high-resolution LC-ESI-MS/MS.
Identification of polyphenols from antiviral Chamaecrista nictitans extract using high-resolution LC-ESI-MS/MS.
Analytical and bioanalytical chemistry (2014-06-28)
María Luisa Mateos-Martín, Elisabet Fuguet, Allan Jiménez-Ardón, Libia Herrero-Uribe, Giselle Tamayo-Castillo, Josep Lluís Torres
ABSTRACT
Chamaecrista nictitans (L) extract possesses antiviral properties; it acts against the herpes simplex virus, and this may be attributed to its constituent phenolics. Here, high-resolution LC-ESI-MS/MS is used to identify the phenolic components of the most potent fraction of the extract. The fraction is a complex mixture rich in oligomeric proanthocyanidins with a high content of monohydroxyphenol moieties ((epi)fisetinidol, (epi)afzelechin and (epi)guibourtinidol) and A-type linkages, uncommon in other proanthocyanidin-rich phenolic extracts, such as those from grape seeds or pine bark. As monohydroxyphenolic structures and A-type linkages have been related to antiviral effects, particularly through the inhibition of late transcription, we suggest that the fraction of C. nictitans extract exerts its action through a particularly effective combination of proanthocyanidins that include these two structural features.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Aciclovir, European Pharmacopoeia (EP) Reference Standard
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Apigenin, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acycloguanosine, ≥99% (HPLC), powder
USP
Acyclovir, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%