MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Protective effects of hydroxytyrosol-supplemented refined olive oil in animal models of acute inflammation and rheumatoid arthritis.
Protective effects of hydroxytyrosol-supplemented refined olive oil in animal models of acute inflammation and rheumatoid arthritis.
The Journal of nutritional biochemistry (2015-01-27)
S Silva, B Sepodes, J Rocha, R Direito, A Fernandes, D Brites, M Freitas, E Fernandes, M R Bronze, M E Figueira
ABSTRACT
Virgin olive oil is the primary source of fat in the Mediterranean diet, and its beneficial health effects have been related with oleic acid and phenolic compounds content. Hydroxytyrosol, a typical virgin olive oil phenolic compound, has beneficial antioxidant and anti-inflammatory properties as previously reported. The aim of this study was to evaluate the effect of hydroxytyrosol-supplemented refined olive oil at 0.5 and 5 mg/kg in a rodent model of rheumatoid arthritis. Rheumatoid arthritis was induced by intradermic administration, in male Wistar rats, of Freund's adjuvant with collagen type II on days 1 and 21. Hydroxytyrosol-supplemented refined olive oils were administrated by gavage from day 23 until day 35. The treatment at 5-mg/kg dose significantly decreased paw edema (P<.01), histological damage, cyclooxygenase-2 and inducible nitric oxide synthase expression, and markedly reduced the degree of bone resorption, soft tissue swelling and osteophyte formation, improving articular function in treated animals. Acute inflammation, induced by carrageenan, was also evaluated for hydroxytyrosol-supplemented refined olive oils at 0.5 and 5 mg/kg. Both doses significantly reduced paw edema (P<.001). Our results suggest that the supplementation of refined olive oil with hydroxytyrosol may be advantageous in rheumatoid arthritis with significant impact not only on chronic inflammation but also on acute inflammatory processes.
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, Molecular Biology, ≥99.5% (KT)
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%