MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Benzidine induces epithelial-mesenchymal transition in human uroepithelial cells through ERK1/2 pathway.
Benzidine induces epithelial-mesenchymal transition in human uroepithelial cells through ERK1/2 pathway.
Biochemical and biophysical research communications (2015-03-12)
Li Zhao, Hao Geng, Zhao-Feng Liang, Zhi-Qiang Zhang, Tao Zhang, De-Xin Yu, Cai-Yun Zhong
ABSTRACT
Prolonged benzidine exposure is a known cause of urothelial carcinoma (UC). Benzidine-induced epithelial-to-mesenchymal transition (EMT) is critically involved in cell malignant transformation. The role of ERK1/2 in regulating benzidine-triggered EMT has not been investigated. This study was to investigate the regulatory role of ERK1/2 in benzidine-induced EMT. By using wound healing and transwell chamber migration assays, we found that benzidine could increase SV-HUC-1 cells invasion activity, western blotting and Immunofluorescence showed that the expression levels of Snail, β-catenin, Vimentin, and MMP-2 were significantly increased, while, the expression levels of E-cadherin, ZO-1 were decreased. To further demonstrate the mechanism in this process, we found that the phosphorylation of ERK1/2, p38, JNK and AP-1 proteins were significantly enhanced compared to the control group (*P < 0.05). Afterward, treated with MAPK pathways inhibitors, only ERK inhibitor(U0126)could reduce the expression of EMT markers in SV-HUC-1 cells, but not p38 and JNK inhibitor(SB203580, SP600125), which indicated that benzidine induces the epithelial-mesenchymal transition in human uroepithelial cells through ERK1/2 pathway. Taken together, findings from this study could provide into the molecular mechanisms by which benzidine exerts its bladder-cancer-promoting effect as well as its target intervention.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material