MilliporeSigma
  • Extraction of functional ingredients from spinach (Spinacia oleracea L.) using liquid solvent and supercritical CO₂ extraction.
Extraction of functional ingredients from spinach (Spinacia oleracea L.) using liquid solvent and supercritical CO₂ extraction.
Journal of the science of food and agriculture (2014-06-17)
Laura Jaime, Erika Vázquez, Tiziana Fornari, María del Carmen López-Hazas, Mónica R García-Risco, Susana Santoyo, Guillermo Reglero
ABSTRACT
In this work three different techniques were applied to extract dry leaves of spinach (Spinacia oleracea): solid-liquid extraction (SLE), pressurised liquid extraction (PLE) and supercritical fluid extraction (SFE) to investigate the influence of extraction solvent and technique on extracts composition and antioxidant activity. Moreover, the influence of carotenoids and phenolic compounds on the antioxidant and anti-inflammatory activities of spinach extracts was also studied. The higher concentrations of carotenoids and the lower content of phenolic compounds were observed in the supercritical CO₂ extracts; whereas water and/or ethanol PLE extracts presented low amounts of carotenoids and the higher concentrations of phenolic compounds. PLE extract with the highest content of phenolic compounds showed the highest antioxidant activity, although SFE carotenoid rich extract also showed a high antioxidant activity. Moreover, both extracts presented an important anti-inflammatory activity. PLE seems to be a good technique for the extraction of antioxidant and anti-inflammatory compounds from spinach leaves. Moreover, spinach phenolic compounds and carotenoids present a high antioxidant activity, whereas spinach carotenoids seem to show a higher anti-inflammatory activity than phenolic compounds. It is worth noting that of our knowledge this is the first time the anti-inflammatory activity of lipophilic extracts from spinach leaves is reported.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Supelco
Triethylamine, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Pricing and availability is not currently available.
Supelco
tert-Butyl methyl ether, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99.5%
Pricing and availability is not currently available.
Supelco
tert-Butyl methyl ether, HPLC grade, suitable for HPLC, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Xanthophyll, from marigold
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl methyl ether, SAJ special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Pricing and availability is not currently available.