MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Adenosine monophosphate-activated protein kinase activation protects against sepsis-induced organ injury and inflammation.
Adenosine monophosphate-activated protein kinase activation protects against sepsis-induced organ injury and inflammation.
The Journal of surgical research (2014-12-03)
Daniel A Escobar, Ana M Botero-Quintero, Benjamin C Kautza, Jason Luciano, Patricia Loughran, Sophie Darwiche, Matthew R Rosengart, Brian S Zuckerbraun, Hernando Gomez
ABSTRACT
Mortality in sepsis is most often attributed to the development of multiple organ failure. In sepsis, inflammation-mediated endothelial activation, defined as a proinflammatory and procoagulant state of the endothelial cells, has been associated with severity of disease. Thus, the objective of this study was to test the hypothesis that adenosine monophosphate-activated protein kinase (AMPK) activation limits inflammation and endothelium activation to protect against organ injury in sepsis. 5-aminoimidazole-4-carboxamide ribonucleotide (AICAR), which is an adenosine monophosphate analog, has been used to upregulate activity of AMPK. Compound C is a cell-permeable pyrrazolopyrimidine compound that inhibits AMPK activity. Wild-type mice underwent cecal ligation and puncture (CLP) or sham surgery. Mice were randomized to vehicle, AICAR, or compound C. Mouse kidney endothelial cells were used for in vitro experiments. Renal and liver function were determined by serum cystatin C, blood urea nitrogen (BUN), creatinine, and alanine aminotransferase. Serum cytokines were measured by enzyme-linked immunosorbent assay. Microvascular injury was determined using Evans blue dye and electron microscopy. Immunohistochemistry was used to measure protein levels of phospho-AMPK (p-AMPK), microtubule-associated protein 1A/1B-light chain 3 (LC3), and intracellular adhesion molecule. LC3 levels were used as a measure of autophagosome formation. AICAR decreased liver and kidney injury induced by CLP and minimized cytokine elevation in vivo and in vitro. CLP increased renal and hepatic phosphorylation of AMPK and autophagic signaling as determined by LC3. Inhibition of AMPK with compound C prevented CLP-induced autophagy and exacerbated tissue injury. Additionally, CLP led to endothelial injury as determined by electron microscopy and Evans blue dye extravasation, and AICAR limited this injury. Furthermore, AICAR limited CLP and lipopolysaccharide (LPS)-induced upregulation of intracellular adhesion molecule in vivo and in vitro and decreased LPS-induced neutrophil adhesion in vitro. In this model, activation of AMPK was protective, and AICAR minimized organ injury by decreasing inflammatory cytokines and endothelial activation. These data suggest that AMPK signaling influences sepsis or LPS-induced endothelial activation and organ injury.
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Dacarbazine impurity B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC