MilliporeSigma
  • Bacteriophage application restores ethanol fermentation characteristics disrupted by Lactobacillus fermentum.
Bacteriophage application restores ethanol fermentation characteristics disrupted by Lactobacillus fermentum.
Biotechnology for biofuels (2015-09-05)
Mei Liu, Kenneth M Bischoff, Jason J Gill, Miranda D Mire-Criscione, Joel D Berry, Ry Young, Elizabeth J Summer
ABSTRACT
Contamination of corn mash by lactic acid bacteria (LAB) reduces the efficiency of the ethanol fermentation process. The industry relies heavily on antibiotics for contamination control and there is a need to develop alternative methods. The goals of this study were to determine the diversity and abundance of bacteria contaminating commercial ethanol fermentations, and to evaluate the potential of anti-LAB bacteriophages in controlling production losses. Bacterial populations in 27 corn mash samples collected from nine different commercial plants were determined by pyrosequencing of 16S rRNA amplicons. The results showed that the most abundant bacteria (>50 % of total population) in 24 of the 27 samples included LAB genera such as Lactobacillus, Streptococcus, Lactococcus, Weissella, Enterococcus, and Pediococcus. Lactobacillus was identified as the most prevalent genus at all fermentation stages in all plants, accounting for between 2.3 and 93.7 % of each population and constituting the major genus (>50 %) in nine samples from five plants and the most abundant genus in five other samples. Lactobacillus species, including L. delbrueckii, L. fermentum, L. mucosae, and L. reuteri were the most well-represented species. Two bacteriophages that target L. fermentum strains from ethanol plants, vB_LfeS_EcoSau and vB_LfeM_EcoInf (EcoSau and EcoInf), were isolated and characterized as a siphophage and a myophage, respectively. Analysis of the 31,703 bp genome of EcoSau revealed its similarity to the P335-like phage group, and the 106,701 bp genome of phage EcoInf was determined to be a novel phage type despite its distant relationship to the SPO1-like phages. Addition of phages EcoSau and EcoInf to L. fermentum-contaminated corn mash fermentation models restored the yields of ethanol and reduced levels of residual glucose, lactic acid, and acetic acid to that comparable to the infection-free control. This study provides detailed insight into the microbiota contaminating commercial ethanol fermentations, and highlights the abundance of LAB, especially L. delbrueckii, L. fermentum, L. mucosae, and L. reuteri, in the process. This study suggests that phages with broad coverage of major LAB species can be applied directly to corn mash for antibiotic-free control of contamination in the ethanol fermentation industry.
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Pricing and availability is not currently available.
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Lactic acid, 88%, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Lactic acid, natural, ≥85%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, 94.8-95.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ammonium sulfate, ≥99.5%
Pricing and availability is not currently available.