MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of chondrocyte-derived extracellular matrix in a dry eye mouse model.
Effects of chondrocyte-derived extracellular matrix in a dry eye mouse model.
Molecular vision (2015-11-26)
Chae Eun Kim, Ha Na Oh, Ji Hyun Lee, Jae Wook Yang
ABSTRACT
The occurrence of repetitive dry eye is accompanied by inflammation. This study investigated the anti-inflammatory effects of chondrocyte-derived extracellular matrix (CDECM) on the cornea and conjunctiva in a dry eye mouse model. Dry eyes were experimentally induced in 12- to 16-week-old NOD.B10.H2(b) mice (Control) via subcutaneous injections of scopolamine (muscarinic receptor blocker) and exposure to an air draft for 10 days (desiccation stress [DS] 10D group). Tear volume and corneal smoothness were measured at 3, 5, 7, and 10 days after the instillation of PBS (PBS group) or CDECM (CDECM group). The corneas and conjunctivas were sectioned and stained with hematoxylin and eosin (H&E) and periodic acid Schiff (PAS). The expression of inflammatory markers (i.e., tumor necrosis factor-α [TNF-α], matrix metalloproteinase-2 [MMP-2], MMP-9, intercellular adhesion molecule-1 [ICAM-1], and vascular cell adhesion molecule-1 [VCAM-1]) was detected by quantitative real-time (qRT)-PCR and western blotting. All data were statistically processed using SPSS version 18.0. The instillation of CDECM after the removal of the DS increased tear production by up to 3.0-fold, and corneal smoothness improved to 80% compared to the PBS group (p<0.05). In the CDECM group, the detachment of the corneal epithelial cells was reduced by 73.3% compared to the PBS group, and the conjunctival goblet cell density was significantly recovered to the control levels (p<0.05). The expression of inflammatory factors was decreased in the cornea and conjunctiva of the CDECM group compared to the PBS group. These observations suggest that CDECM induced effective anti-inflammatory improvements in the cornea and conjunctiva in this experimental model of dry eye.
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Scopolamine hydrobromide, meets USP testing specifications
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%