MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Participation of autophagy in the cytotoxicity against breast cancer cells by cisplatin.
Participation of autophagy in the cytotoxicity against breast cancer cells by cisplatin.
Oncology reports (2015-05-26)
Meng Shen, Wei-Ming Duan, Meng-Yao Wu, Wen-Jie Wang, Lu Liu, Meng-Dan Xu, Jie Zhu, Dao-Ming Li, Qi Gui, Lian Lian, Fei-Ran Gong, Kai Chen, Wei Li, Min Tao
ABSTRACT
Breast cancer is one of the most common cancers affecting women worldwide. Conventional chemotherapy is still one of the major approaches to the treatment of breast cancer. Autophagy, also termed as type II programmed cell death (PCD), exhibits either a protumorigenic or antitumorigenic function. In the present study, we investigated whether autophagy could be involved in the effect of chemotherapy against breast cancer. Epirubicin, docetaxel, methotrexate, cyclophosphamide, fluorouracil (5-FU) and cisplatin were applied in the present investigation. All of these chemotherapeutics presented cytotoxicity against breast cancer cells. DsRed-LC3 reporter assay revealed that only docetaxel and cisplatin induced autophagy. Autophagy inhibitor 3-methyladenine (3-MA) strengthened the cytotoxicity of docetaxel, yet impaired the cytotoxicity of cisplatin, suggesting that docetaxel stimulates protumorigenic autophagy, while cisplatin-induced autophagy could be antitumorigenic. Real-time PCR revealed that cisplatin upregulated multiple autophagy-related genes, including AMBRA1, ATG3, ATG4C, ATG4D, ATG5, ATG7, ATG13, ATG14, ATG16L2, Beclin1, DRAM1, GABARAP, GABARAPL1, GABARAPL2, HDAC6, IRGM, MAP1LC3B and ULK1, indicating that cisplatin induced autophagy through a multiple mechanism involved manner.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor