MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The Nitric oxide/CGMP/KATP pathway mediates systemic and central antinociception induced by resistance exercise in rats.
The Nitric oxide/CGMP/KATP pathway mediates systemic and central antinociception induced by resistance exercise in rats.
The International journal of neuroscience (2014-10-02)
Giovane S Galdino, Carlos H Xavier, Renato Almeida, Grazielle Silva, Marcos A Fontes, Gustavo Menezes, Igor D Duarte, Andrea C Perez
ABSTRACT
Resistance exercise (RE) is characterized to increase strength, tone, mass, and/or muscular endurance and also for produces many beneficial effects, such as blood pressure and osteoporosis reduction, diabetes mellitus control, and analgesia. However, few studies have investigated endogenous mechanisms involved in the RE-induced analgesia. Thus, the aim of this study was evaluate the role of the NO/CGMP/KATP pathway in the antinociception induced by RE. Wistar rats were submitted to acute RE in a weight-lifting model. The nociceptive threshold was measured by mechanical nociceptive test (paw-withdrawal). To investigate the involvement of the NO/CGMP/KATP pathway the following nitric oxide synthase (NOS) non-specific and specific inhibitors were used: N-nitro-l-arginine (NOArg), Aminoguanidine, N5-(1-Iminoethyl)-l-ornithine dihydrocloride (l-NIO), Nω-Propyl-l-arginine (l-NPA); guanylyl cyclase inhibitor, 1H-[1,2,4]oxidiazolo[4,3-a]quinoxalin-1-one (ODQ); and KATP channel blocker, Glybenclamide; all administered subcutaneously, intrathecally and intracerebroventricularly. Plasma and cerebrospinal fluid (CSF) nitrite levels were determined by spectrophotometry. The RE protocol produced antinociception, which was significantly reversed by NOS specific and unspecific inhibitors, guanylyl cyclase inhibitor (ODQ) and KATP channel blocker (Glybenclamide). RE was also responsible for increasing nitrite levels in both plasma and CSF. These finding suggest that the NO/CGMP/KATP pathway participates in antinociception induced by RE.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture