MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Functional analyses of polymorphic variants of human terminal deoxynucleotidyl transferase.
Functional analyses of polymorphic variants of human terminal deoxynucleotidyl transferase.
Genes and immunity (2015-06-05)
A Troshchynsky, I Dzneladze, L Chen, Y Sheng, V Saridakis, G E Wu
ABSTRACT
Human terminal deoxynucleotidyl transferase (hTdT) is a DNA polymerase that functions to generate diversity in the adaptive immune system. Here, we focus on the function of naturally occurring single-nucleotide polymorphisms (SNPs) of hTdT to evaluate their role in genetic-generated immune variation. The data demonstrate that the genetic variations generated by the hTdT SNPs will vary the human immune repertoire and thus its responses. Human TdT catalyzes template-independent addition of nucleotides (N-additions) during coding joint formation in V(D)J recombination. Its activity is crucial to the diversity of the antigen receptors of B and T lymphocytes. We used in vitro polymerase assays and in vivo human cell V(D)J recombination assays to evaluate the activity and the N-addition levels of six natural (SNP) variants of hTdT. In vitro, the variants differed from wild-type hTdT in polymerization ability with four having significantly lower activity. In vivo, the presence of TdT varied both the efficiency of recombination and N-addition, with two variants generating coding joints with significantly fewer N-additions. Although likely heterozygous, individuals possessing these genetic changes may have less diverse B- and T-cell receptors that would particularly effect individuals prone to adaptive immune disorders, including autoimmunity.
Sigma-Aldrich
Potassium acetate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Boric acid, SAJ first grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
SAFC
L-Glutamine
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Boric acid, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, BioReagent, Molecular Biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)