MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Members of the Plant CRK Superfamily Are Capable of Trans- and Autophosphorylation of Tyrosine Residues.
Members of the Plant CRK Superfamily Are Capable of Trans- and Autophosphorylation of Tyrosine Residues.
The Journal of biological chemistry (2015-05-15)
Keiichirou Nemoto, Nobuaki Takemori, Motoaki Seki, Kazuo Shinozaki, Tatsuya Sawasaki
ABSTRACT
Protein phosphorylation on Tyr residues is a key post-translational modification in mammals. In plants, recent studies have identified Tyr-specific protein phosphatase and Tyr-phosphorylated proteins in Arabidopsis by phosphoproteomic screenings, implying that plants have a Tyr phosphorylation signal pathway. However, little is known about the protein kinases (PKs) involved in Tyr phosphorylation in plants. Here, we demonstrate that Arabidopsis calcium-dependent protein kinase (CDPK/CPK)-related PKs (CRKs) have high Tyr-autophosphorylation activity and that they can phosphorylate Tyr residue(s) on substrate proteins in Arabidopsis. To identify PKs for Tyr phosphorylation, we examined the autophosphorylation activity of 759 PKs using an Arabidopsis protein array based on a wheat cell-free system. In total, we identified 38 PKs with Tyr-autophosphorylation activity. The CRK family was a major protein family identified. A cell-free substrate screening revealed that these CRKs phosphorylate β-tubulin (TBB) 2, TBB7, and certain transcription factors (TFs) such as ethylene response factor 13 (ERF13). All five CRKs tested showed Tyr-auto/trans-phosphorylation activity and especially two CRKs, CRK2 and CRK3, showed a high ERF13 Tyr-phosphorylation activity. A cell-based transient expression assay revealed that Tyr(16/)Tyr(207) sites in ERF13 were phosphorylated by CRK3 and that Tyr phosphorylation of endogenous TBBs occurs in CRK2 overexpressing cells. Furthermore, crk2 and crk3 mutants showed a decrease in the Tyr phosphorylation level of TBBs. These results suggest that CRKs have Tyr kinase activity, and these might be one of the major PKs responsible for protein Tyr phosphorylation in Arabidopsis plants.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acrylamide, ≥98.0%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
TBB, ≥98% (HPLC), solid
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%